missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531993
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1. Predicted reactivity: Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
VSIG1 | |
Polyclonal | |
Unconjugated | |
VSIG1 | |
1700062D20Rik; 4930405J24Rik; cell surface A33 antigen; CHT1; ChT1 thymocyte antigen; dJ889N15.1; Glycoprotein A34; GPA34; MGC44287; V-set and immunoglobulin domain containing 1; V-set and immunoglobulin domain-containing protein 1; Vsig1; VSIG1 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340547 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.74 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q86XK7 | |
VSIG1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 212 of Human VSIG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |