missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS53 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520549
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is 293 cell lysate. PA5-20549 can be used with blocking peptide PEP-0669.
The sorting of acid hydrolases to lysosomes rely on mannose 6-phosphate receptors that cycle between the trans-Golgi network (TGN) and endosomes. The maintenance of this cycle requires the function of the mammalian Golgi-associated retrograde protein (GARP) complex which is composed of three subunits: VPS52, VPS53, and VPS54. Depletion of any of these three proteins, such as by RNAi, impairs the retrograde transport of multiple TGN proteins. VPS53 was identified as an HIV dependency factor (HDF) and plays a role in viral entry to the cell, suggesting that VPS53 may be an important drug target in HIV treatment. At least five isoforms of VPS53 are known to exist.Specifications
VPS53 | |
Polyclonal | |
Unconjugated | |
Vps53 | |
2010002A08Rik; 2310040I21Rik; 3100002B05Rik; Are1; Hccs1; hVps53L; PCH2E; pp13624; RGD1311391; SAC2 suppressor of actin mutations 2-like protein; Sacm2l; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting 53 (yeast); vacuolar protein sorting 53 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; Vacuolar protein sorting-associated protein 53 homolog; Vps52; Vps53; VPS53 GARP complex subunit; VPS53, GARP complex subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
287535, 55275, 68299 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5VIR6, Q8CCB4 | |
Vps53 | |
An 18 amino acid peptide from near the center of human VPS53. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |