missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS45 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSV00017G
This item is not returnable.
View return policy
Description
Glycerol (1:1) may be added for additional stability.
expressed in brain and testis; may play a role in intracellular vesicle mediated transport.
Specifications
| Vps45 | |
| Polyclonal | |
| Unconjugated | |
| P97390, O08700 | |
| VPS45 | |
| A synthetic peptide from rat VPS45 conjugated to an immunogenic carrier protein was used as the antigen | |
| 500 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Vps45 | |
| Vps45, VPS45, rvps45, Vps45a | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 22365, 64516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction