missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vomeronasal 1 Receptor A1/A13/A14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSR00230W
This item is not returnable.
View return policy
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor; may play a role in olfactory sensory perception. Putative pheromone receptor implicated in the regulation of social as well as reproductive behavior. Expressed in 1-4% of neurons of the vomeronasal organ. Only one pheromone receptor gene may be expressed in a particular neuron. Not expressed in the main olfactory epithelium.
Specifications
| Vomeronasal 1 Receptor A1/A13/A14 | |
| Polyclonal | |
| Vom1r94 | |
| V1ra14, Vom1r105, Vom1r94, Vomeronasal type-1 receptor A1, A13, A14, vomeronasal receptor 1, pheromone receptor VN1, Vnr1, V1A13, V1ra13, V1A14, V1ra14 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q5J3K9, Q5J3M4, Q62850 | |
| V1RA14, VOM1R105, VOM1R94 | |
| Synthetic peptide from rat vomeronasal type-1 receptor A1, A13, A14. | |
| RUO | |
| 286893, 297442, 494265 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction