missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitronectin Mouse anti-Human, Clone: 10E12, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0205
This item is not returnable.
View return policy
Description
A suggested positive control for this product is human plasma.
The protein encoded by this gene is a member of the pexin family. It is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. It is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
Specifications
| Vitronectin | |
| Monoclonal | |
| Unconjugated | |
| P04004 | |
| VTN | |
| Protein purified from Human plasma | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 10E12 | |
| VTN | |
| V75, VN, VNT | |
| Mouse | |
| Protein G | |
| RUO | |
| 7448 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction