Learn More
VSNL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530258
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: IMR32, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (98%), Chicken (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.Specifications
VSNL1 | |
Polyclonal | |
Unconjugated | |
VSNL1 | |
21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24877, 26950, 7447 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P62760, P62761, P62762 | |
VSNL1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 167 of Human Visinin-like 1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |