Learn More
vasoactive intestinal peptide receptor 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant VIPR2.
Brand: Abnova H00007434-A01.50uL
Description
Vasoactive intestinal peptide (VIP; MIM 192320) and pituitary adenylate cyclase activating polypeptide (PACAP; MIM 102980) are homologous peptides that function as neurotransmitters and neuroendocrine hormones. While the receptors for VIP and PACAP share homology, they differ in their substrate specificities and expression patterns. See VIPR1 (MIM 192321) and ADCYAP1R1(MIM 102981).[supplied by OMIM
Sequence: ECRFHLEIQEEETKCAELLRSQTEKHKACSGVWDNITCWRPANVGETVTVPCPKVFSNFYSKAGNISKNCTSDGWSETFPDFVDACGYSDPEDESKITFYILVSpecifications
| vasoactive intestinal peptide receptor 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant VIPR2. | |
| VIPR2 | |
| FLJ16511/VPAC2/VPCAP2R | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003382 | |
| VIPR2 | |
| VIPR2 (NP_003373, 24 a.a. ∼ 126 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 7434 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.