missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vinexin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530361
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The SORBS3 gene encodes 2 isoforms vinexin-alpha and -beta that are vinculin (VCL; MIM 193065)-binding cytoskeletal proteins involved in focal adhesion and cell-cell adhesion (Kioka et al., 1999 [PubMed 9885244]).
Specifications
| Vinexin | |
| Polyclonal | |
| Unconjugated | |
| SORBS3 | |
| Scam1; SCAM-1; SH3 domain protein 4; SH3 domain-containing protein SH3P3; SH3-containing adapter molecule 1; Sh3d4; SH3P3; sorbin and SH3 domain containing 3; sorbin and SH3 domain-containing protein 3; Sorbs3; Vinexin; vinexin alpha; vinexin beta; vinexin beta (SH3-containing adaptor molecule-1); vinexin-g | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10174, 20410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O60504, Q9R1Z8 | |
| SORBS3 | |
| Recombinant protein encompassing a sequence within the center region of human Vinexin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction