Learn More
Vimentin Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA116759
Code : LD
Additional Details : Weight : 0.01000kg
Description
Vimentin Polyclonal antibody specifically detects Vimentin in Bovine, Canine, Chicken, Equine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Vimentin | |
Polyclonal | |
Unconjugated | |
Vim | |
cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
Chicken | |
IgY fraction | |
RUO | |
100056088, 100522394, 22352, 280955, 420519, 477991, 7431, 81818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
16.6 mg/mL | |
PBS with 5mM sodium azide | |
P02543, P08670, P09654, P20152, P31000, P48616 | |
Vim | |
The immunogen used to generate our antibody was recombinant vimentin purified from E. coli. | |
100 μL | |
Primary | |
Bovine, Canine, Chicken, Equine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgY |