Learn More
VENT homeobox homolog (Xenopus laevis), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human VENTX protein.
Brand: Abnova H00027287-B01P.50ug
Description
This gene encodes a member of the Vent family of homeodomain proteins. The encoded protein may function as a transcriptional repressor and be involved in mesodermal patterning and hemopoietic stem cell maintenance. Multiple pseudogenes exist for this gene. A transcribed pseudogene located on chromosome X may lead to antigen production in certain melanomas. [provided by RefSeq
Sequence: MRLSSSPPRGPQQLSSFGSVDWLSQSSCSGPTHTPRPADFSLGSLPGPGQTSGAREPPQAVSIKEAAGSSNLPAPERTMAGLSKEPNTLRAPRVRTAFTMEQVRTLEGVFQHHQYLSPLERKRLAREMQLSEVQIKTWFQNRRMKHKRQMQDPQLHSPFSGSLHAPPAFYSTSSGLANGLQLLCPWAPLSGPQALMLPPGSFWGLCQVAQEALASAGASCCGQPLASHPPTPGRPSLGPALSTGPRGLCAMPQTGDAFSpecifications
| VENT homeobox homolog (Xenopus laevis) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human VENTX protein. | |
| VENTX | |
| HPX42B/MGC119910/MGC119911/NA88A/VENTX2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 27287 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014468.2 | |
| VENTX | |
| VENTX (NP_055283.1, 1 a.a. ∼ 258 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.