missing translation for 'onlineSavingsMsg'
Learn More

VASP Mouse, Unlabeled, Clone: 43, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610448

 View more versions of this product

Product Code. 15835469

  • ONLINE EXCLUSIVE PRICE
    £297.00
    £284.36 / 1 set
    Save £12.64
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Vasodilator-stimulated phosphoprotein (VASP), a substrate for cAMP- and cGMP-dependent kinases, is associated with actin filaments, focal adhesions, and dynamic membrane regions. VASP is composed of several distinct domains: a central L-proline-rich domain contains a GPPPPP motif as a single copy and as a three-fold tandem repeat, as well as three conserved phosphorylation sites for cyclic nucleotide-dependent protein kinases (Ser157, Ser239, and Thr278). A C-terminal domain contains a repetitive mixed-charge cluster which is predicted to form an α-helix. The C-terminal domain appears to be responsible for anchoring at focal adhesion sites. VASP has been shown to be a ligand for profilins. Profilins bind to the poly-L-proline motifs of VASP and it is postulated that these two molecules act in concert to convey signal transduction to actin filament formation.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

VASP
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Human
IgG1
Western Blot
43
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.