missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VASN Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547258
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
VASN Polyclonal antibody specifically detects VASN in Human samples. It is validated for Western Blot.Specifications
VASN | |
Polyclonal | |
Unconjugated | |
VASN | |
2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin | |
Goat | |
Antigen affinity chromatography | |
RUO | |
114990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6EMK4 | |
VASN | |
CHO-derived recombinant human Vasorin extracellular domain | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |