Learn More
VAMP (vesicle-associated membrane protein)-associated protein B and C (D01P), Rabbit anti-Human, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human VAPB protein.
Brand: Abnova H00009217-D01P.100ug
Description
The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking. [provided by RefSeq
Sequence: MAKVEQVLSLEPQHELKFRGPFTDVVTTNLKLGNPTDRNVCFKVKTTAPRRYCVRPNSGIIDAGASINVSVMLQPFDYDPNEKSKHKFMVQSMFAPTDTSDMEAVWKEAKPEDLMDSKLRCVFELPAENDKPHDVEINKIISTTASKTETPIVSKSLSSSLDDTEVKKVMEECKRLQGEVQRLREENKQFKEEDGLRMRKTVQSNSPISALAPTGKEEGLSTRLLALVVLFFIVGVIIGKIALSpecifications
| VAMP (vesicle-associated membrane protein)-associated protein B and C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human VAPB protein. | |
| VAPB | |
| ALS8/VAMP-B/VAMP-C/VAP-B/VAP-C | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9217 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004738.3 | |
| VAPB | |
| VAPB (NP_004729.1, 1 a.a. ∼ 243 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.