Learn More
Vβ 8 T-Cell Receptor Mouse anti-Mouse, Unlabeled, Clone: F23.1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553859
Description
The F23.1 antibody reacts with the Vβ 8.1, Vβ 8.2, and Vβ 8.3 T-cell receptors (TCR) of mice having the b haplotype (e.g., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V8 subfamily gene loci are deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII) haplotype. Vβ 8.1 TCR-bearing T lymphocytes are clonally eliminated in mice expressing superantigen coded by Mtv-7 (Mls-1a, Mlsa) provirus (e.g., AKR, CBA/J, C58, DBA/2), and activation or elimination of Vβ 8.1 TCR-expressing T cells by this determinant is partially dependent upon presentation by I-E. Mtv-43 and/or exogenous MMTV-SW superantigens also cause incomplete elimination of Vβ 8.1 TCR-bearing T cells. In addition to expression on conventional T lymphocytes, Vβ 8.2 is the predominant β chain of the TCR on NK-T cells. Staphylococcal enterotoxin B, in association with antigen-presenting cells expressing I-A and/or I-E, stimulates lymphocytes bearing Vβ 8 TCR and selectively eliminates those T cells in vivo. Soluble and plate-bound F23.1 antibody activates Vβ 8 TCR-bearing T cells, soluble antibody blocks cytolysis mediated by Vβ 8 TCR-bearing cytotoxic T lymphocytes, and in vivo treatment of neonatal mice can arrest intrathymic maturation of Vβ 8 TCR-bearing T cells.
Host Species: Mouse
Clone: F23.1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB.B Mouse Lymph-Node and Spleen T Cells
ELISA, Flow Cytometry, Immunoprecipitation, Western Blotting
Specifications
| Vβ 8 T-Cell Receptor | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB.B Mouse Lymph-Node and Spleen T Cells | |
| 0.25 mg | |
| Immunology | |
| Bovine, Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| F23.1 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.