Learn More
Vδ2 TCR Mouse anti-Human, FITC, Clone: B6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555738
Description
Reacts with the human variable δ2 subunit (Vδ2) of γ/δ T cell receptor (TCR). The γ/δ TCR is composed of two disulfide linked glycoproteins. The delta chain is approximately 40-60kDa and is restricted to peripheral blood T cells and thymocytes. The majority of normal peripheral blood γ/δ T cells express a Vγ/9[+]/ Vδ2[+] phenotype. The reason for this selection in the T-cell repertoire is not well understood.
Host Species: Mouse
Clone: B6
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| Vδ2 TCR | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| B6 | |
| FITC | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.