Learn More
Vβ 14 TCR Rat anti-Mouse, Biotin, Clone: 14-2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553257
Description
The 14-2 antibody reacts with the Vβ 14 T-Cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL, SWR), b (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c (e.g., RIII) halpotypes of the Tcrb gene complex. Vβ 14 TCR-
Host Species: Rat
Clone: 14-2
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: Mouse B10.A Helper T-Cell Clone J9.19
Flow Cytometry
Specifications
| Vβ 14 TCR | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse B10.A Helper T-Cell Clone J9.19 | |
| 0.25 mg | |
| Immunology | |
| Mouse | |
| IgM κ |
| Flow Cytometry | |
| 14-2 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.