Learn More
upstream transcription factor 1, Mouse, Clone: 2A7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant USF1.
Brand: Abnova H00007391-M02.100ug
Description
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MKGQQKTAETEEGTVQIQEGAVATGEDPTSVAIASIQSAATFPDPNVKYVFRTENGGQVMYRVIQVSEGQLDGQTEGTGAISGYPATQSMTQAVIQGAFTSDDAVDTEGTSpecifications
| upstream transcription factor 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007122 | |
| USF1 | |
| USF1 (NP_009053, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2A7 | |
| Mouse monoclonal antibody raised against a partial recombinant USF1. | |
| USF1 | |
| FCHL/FCHL1/HYPLIP1/MLTF/MLTFI/UEF/bHLHb11 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 7391 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.