Learn More
ULK2, Mouse, Clone: 7G5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ULK2.
Brand: Abnova H00009706-M07.100ug
Description
This gene encodes a protein that is similar to a serine/threonine kinase in C. elegans which is involved in axonal elongation. The structure of this protein is similar to the C. elegans protein in that both proteins have an N-terminal kinase domain, a central proline/serine rich (PS) domain, and a C-terminal (C) domain. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq
Sequence: FLRTRTTSVGPSNSGGSLCAMSGRVCVGSPPGPGFGSSPPGAEAAPSLRYVPYGASPPSLEGLITFEAPELPEETLMEREHTDTLRHLNVMLMFTECVLDLSpecifications
| ULK2, | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC034988 | |
| ULK2 | |
| ULK2 (AAH34988, 743 a.a. ∼ 843 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 κ |
| ELISA, Western Blot | |
| 7G5 | |
| Mouse monoclonal antibody raised against a partial recombinant ULK2. | |
| ULK2 | |
| KIAA0623/Unc51.2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9706 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.