Learn More
ubiquitin-conjugating enzyme E2Q family member 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human UBE2Q1 protein.
Brand: Abnova H00055585-B01.50uL
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is 98% identical to the mouse counterpart. [provided by RefSeq
Sequence: MELLTKQGWSSAYSIESVIMQISATLVKGKARVQFGANKSQYSLTRAQQSYKSLVQIHEKNGWYTPPKEDGSpecifications
| ubiquitin-conjugating enzyme E2Q family member 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human UBE2Q1 protein. | |
| UBE2Q1 | |
| GTAP/NICE-5/PRO3094/UBE2Q | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC015316.1 | |
| UBE2Q1 | |
| UBE2Q1 (AAH15316.1, 1 a.a. ∼ 71 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 55585 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.