Learn More
anti-Tyrosine Hydroxylase, Polyclonal PA5-29617, Invitrogen™
Specifically detect Tyrosine Hydroxylase in Human, Mouse, Rat samples using Thermo Scientifc™ Pierce™ anti-Tyrosine Hydroxylase Polyclonal, validated for ICC, IHC (P) and WB
Brand: Thermo Scientific PA529617
Description
Specifically detect Tyrosine Hydroxylase in Human, Mouse, Rat samples using Thermo Scientifc™ Pierce™ anti-Tyrosine Hydroxylase Polyclonal, validated for ICC, IHC (P) and WB
The protein encoded by this gene is involved in the conversion of tyrosine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines, hence plays a key role in the physiology of adrenergic neurons. Mutations in this gene have been associated with autosomal recessive Segawa syndrome. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Host Species:Rabbit
Isotype:IgG
Species Reactivity:Human, Rat
Immunogen:Recombinant fragment corresponding to a region within amino acids 150 and 490 of Human Tyrosine HydroxylaseRecommended for:Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
| Tyrosine Hydroxylase | |
| Polyclonal | |
| P07101 | |
| Affinity Purified | |
| Primary | |
| Store at -20°C. Avoid freeze/Thaw cycles. |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| TH | |
| 100 μL | |
| Human, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.