missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXK tyrosine kinase, Mouse, Clone: 2D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TXK.
Brand: Abnova H00007294-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LIPSNYVTENKITNLEIYEWYHRNITRNQAEHLLRQESKEGAFIVRDSRHLGSYTISVFMGARRSTEAAIKHYQIKKNDSGQWYVAERHAFQSIPELIWYSpecifications
| TXK tyrosine kinase | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003328 | |
| TXK | |
| TXK (NP_003319, 131 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| 7294 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 2D8 | |
| Mouse monoclonal antibody raised against a partial recombinant TXK. | |
| TXK | |
| BTKL/MGC22473/PSCTK5/PTK4/RLK/TKL | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction