Learn More
twist homolog 1 (Drosophila), Mouse, Clone: 2F8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TWIST1.
Brand: Abnova H00007291-M03.100ug
Description
Basic helix-loop-helix (bHLH) transcription factors have been implicated in cell lineage determination and differentiation. The protein encoded by this gene is a bHLH transcription factor and shares similarity with another bHLH transcription factor, Dermo1. The strongest expression of this mRNA is in placental tissue; in adults, mesodermally derived tissues express this mRNA preferentially. Mutations in this gene have been found in patients with Saethre-Chotzen syndrome. [provided by RefSeq
Sequence: PQSYEELQTQRVMANVRERQRTQSLNEAFAALRKIIPTLPSDKLSKIQTLKLAARYIDFLYQVLQSDELDSKMASCSYVAHERLSYAFSVWRMEGAWSMSASHSpecifications
| twist homolog 1 (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000474 | |
| TWIST1 | |
| TWIST1 (NP_000465, 100 a.a. ∼ 202 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2F8 | |
| Mouse monoclonal antibody raised against a partial recombinant TWIST1. | |
| TWIST1 | |
| ACS3/BPES2/BPES3/SCS/TWIST/bHLHa38 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 7291 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.