Learn More
anti-Tubulin-Microtubule Marker, Clone: YL1/2, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB6160-100ug
Description
Host Species: Rat
Clone: YL1/2
Isotype: IgG2a
Cross Reactivity: Mouse, Human, Pig, Saccharomyces cerevisiae, Xenopus laevis, Caenorhabditis elegans, Fruit fly (Drosophila melanogaster), Schizosaccharomyces pombe Predicted to work with a wide range of other species, all Mammals
Immunogen: Full length native protein (purified) (S. cerevisiae).
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
| Tubulin-Microtubule Marker | |
| YL1/2 | |
| Unconjugated | |
| Full length native protein (purified) (S. cerevisiae). | |
| Isotype Loading Control, Signal Transduction, Tags And Cell Markers | |
| Drosophila, Human, Mammalia, Mouse, Pig, S. cerevisiae, S. pombe, Xenopus species |
| Monoclonal | |
| 1mg/mL | |
| Rat | |
| 100 μg | |
| Primary | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.