missing translation for 'onlineSavingsMsg'
Learn More

tissue specific transplantation antigen P35B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human TSTA3 protein.

Brand:  Abnova H00007264-D01P.100ug

Product Code. 16177025

  • £316.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Tissue specific transplantation antigen P35B is a NADP(H)-binding protein. It catalyze the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-deoxymannose to GDP-L-fucose. GDP-L-fucose is the substrate of several fucosyltransferases involved in the expression of many glycoconjugates, including blood group ABH antigens and developmental adhesion antigens. Mutations in this gene may cause leukocyte adhesion deficiency, type II. [provided by RefSeq

Sequence: MGEPQGSMRILVTGGSGLVGKAIQKVVADGAGLPGEDWVFVSSKDADLTDTAQTRALFEKVQPTHVIHLAAMVGGLFRNIKYNLDFWRKNVHMNDNVLHSAFEVGARKVVSCLSTCIFPDKTTYPIDETMIHNGPPHNSNFGYSYAKRMIDVQNRAYFQQYGCTFTAVIPTNVFGPHDNFNIEDGHVLPGLIHKVHLAKSSGSALTVWGTGNPRRQFIYSLDLAQLFIWVLREYNEVEPIILSVGEEDEVSIKEAAEAVVEAMDFHGEVTFDTTKSDGQFKKTASNSKLRTYLPDFRFTPFKQAVKETCAWFTDNYEQARK
Specifications

Specifications

tissue specific transplantation antigen P35B
Polyclonal
Rabbit polyclonal antibody raised against a full-length human TSTA3 protein.
TSTA3
FX/P35B/SDR4E1
Rabbit
Affinity Purified
RUO
7264
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_003313.2
TSTA3
TSTA3 (NP_003304.1, 1 a.a. ∼ 321 a.a) full-length human protein.
100 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.