missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSSK4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530886
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (85%), Pig (87%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).
Specifications
| TSSK4 | |
| Polyclonal | |
| Unconjugated | |
| TSSK4 | |
| 1700020B19Rik; 4933424F08Rik; C14orf20; serine/threonine kinase 22E; serine/threonine-protein kinase 22E; STK22E; testis specific serine kinase 4; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; testis-specific serine/threonine-protein kinase 4; TSK4; TSK-4; Tssk4; TSSK-4; TSSK5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283629 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6SA08 | |
| TSSK4 | |
| Recombinant protein encompassing a sequence within the center region of human TSSK4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction