missing translation for 'onlineSavingsMsg'
Learn More

TSG101 [Tumor Susceptibility Gene 101] Mouse, Unlabeled, Clone: 51, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612697

 View more versions of this product

Product Code. 15896259

  • ONLINE EXCLUSIVE PRICE
    £303.00
    £290.95 / Each
    Save £12.05
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Tumor Susceptibility Gene 101 (TSG101) was identified in a random mutagenesis screen for potential tumor suppressors in NIH 3T3 cells. Altered transcripts of this gene have been detected in sporadic breast cancers and many other human malignancies. Though the function of TSG101 is not clearly understood, its protein structure includes motifs involved in transcription regulation, and TSG101 has been shown to modulate the activation of steroid hormone receptors. In addition, TSG101 may have a role in regulating ubiquination. The N-terminal region of TSG101 contains a domain that resembles the catalytically active region of ubiquitin conjugases. However, TSG101 lacks an active-site cysteine crucial to the function of these conjugases. Interestingly, TSG101 interferes with MDM2 ubiquination leading to a decrease in MDM2 decay and down-regulation of p53 protein. Thus, TSG101 may function as a dominant-negative inhibitor of ubiquination in pathways where protein expression is tightly regulated.

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG101 [Tumor Susceptibility Gene 101]
Monoclonal
250μg/mL
Tumor Susceptibility Gene 101
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
51
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human TSG101 aa. 229-319
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.