Learn More
anti-TSC22D3 Matched Antibody Pair, Ab-1, Abnova
Brand: Abnova H00001831-AP11.1Set
Includes: Rabbit affinity purified polyclonal anti-TSC22D3 (100μg); Mouse monoclonal anti-TSC22D3, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
The protein encoded by this gene shares significant sequence identity with the murine TSC-22 and Drosophila shs, both of which are leucine zipper proteins, that function as transcriptional regulators. The expression of this gene is stimulated by glucocorticoids and interleukin 10, and it appears to play a key role in the anti-inflammatory and immunosuppressive effects of this steroid and chemokine. Transcript variants encoding different isoforms have been identified for this gene.
ELISA
Specifications
| TSC22D3 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TSC22D3. | |
| DIP/DKFZp313A1123/DSIPI/GILZ/TSC-22R/hDIP | |
| Mouse, Rabbit | |
| 1 Set | |
| Transcription Regulation | |
| 1831 | |
| Human | |
| IgG2a |
| ELISA | |
| Unconjugated | |
| TSC22D3 | |
| TSC22D3 | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TSC22D3 (100 μg) 2. Detection antibody: mouse monoclonal anti-TSC22D3, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.