Learn More
tuberous sclerosis 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TSC2.
Brand: Abnova H00007249-A01.50uL
Description
Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: SPPPELEERDVAAYSASLEDVKTAVLGLLVILQTKLYTLPASHATRVYEMLVSHIQLHYKHSYTLPIASSIRLQAFDFLFLLRADSLHRLGLPNKDGVVRFSPYCVCDYMEPERGSEKKSpecifications
| tuberous sclerosis 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TSC2. | |
| TSC2 | |
| FLJ43106/LAM/TSC4 | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| 7249 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000548 | |
| TSC2 | |
| TSC2 (NP_000539, 540 a.a. ∼ 658 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.