Learn More
Trypsin 2 Monoclonal Antibody (410322), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524148
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody does not cross-react with recombinant human (rh) Trypsin 1 or rhTrypsin 3. This antibody recognizes both the pro and mature forms of rhTrypsin 2. Reconstitute at 0.5 mg/mL in sterile PBS.
Trypsin 2 Monoclonal antibody specifically detects Trypsin 2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Trypsin 2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P07478 | |
PRSS2 | |
Mouse myeloma cell line NS0-derived recombinant human Trypsin 2/PRSS2 Ala16-Ser247 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
410322 | |
Unconjugated | |
PRSS2 | |
anionic trypsin; anionic trypsin II; Anionic trypsin-2; anionic trypsinogen; EC 3.4.21; EC 3.4.21.4; MGC111183; MGC120174; MGC120175; pancreatic anionic trypsinogen; pancreatic trypsin 2; pancreatic trypsin II; pretrypsinogen II; protease serine 2 preproprotein; protease, serine 2; protease, serine, 2; protease, serine, 2 (trypsin 2); PRSS2; Ptryss2; Serine protease 2; Ta; Tesp4; Try2; TRY8; TRYP; TRYP2; TRYP8; trypsin 2; trypsin II; trypsin Trypsin II; Trypsin-2; trypsinogen 2; trypsinogen 20; trypsinogen anionic precursor | |
Mouse | |
Protein A/G | |
RUO | |
5645 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |