missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OST00143G
This item is not returnable.
View return policy
Description
Glycerol (1:1) may be added for additional stability. Reconstitute with 500 μL of distilled water. Centrifuge to remove insoluble material.
Specifications
| TRPG | |
| Polyclonal | |
| Unconjugated | |
| Q9VJJ7 | |
| TRPGAMMA | |
| A synthetic peptide from the 4th cytoplasmic loop of drosophila TRPG (Transient receptor potential-gamma protein, TRP gamma) conjugated to an immunogenic carrier protein was used as the antigen | |
| 500 μg | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Trpgamma | |
| TRPG, Transient receptor potential-gamma protein, TRPgamma | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 34991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction