Learn More
TRPM8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529382
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HepG2, Raji. Predicted reactivity: Mouse (90%), Rat (89%), Dog (95%), Bovine (95%), Guinea pig (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRPM8 Polyclonal antibody specifically detects TRPM8 in Human samples. It is validated for Western Blot.Specifications
TRPM8 | |
Polyclonal | |
Unconjugated | |
TRPM8 | |
CMR1; Cold menthol receptor 1; cold/menthol receptor 1; long transient receptor potential channel 6; LTRPC6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trpm8; Trpp8; trp-p8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.56 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q7Z2W7 | |
TRPM8 | |
Recombinant fragment corresponding to a region within amino acids 321 and 611 of Human Trp-p8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |