Learn More
TROP-2 Mouse anti-Human, Brilliant Violet 421, Clone: 162-46, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563243
Description
The 162-46 monoclonal antibody recognizes human Trop-2 which is also known as GA733-1. Trop-2 is encoded by TACSTD2 (tumor-associated calcium signal transducer 2). It is a single-pass type-1 transmembrane glycoprotein. Trop-2 was originally identified to be expressed on human trophoblast and choriocarcinoma cell lines. Trop-2 is expressed in most human carcinomas. Trop-2 appears to be involved in the transduction of an intracellular calcium signal, although the exact physiological mechanism has yet to be elucidated.
Specifications
| TROP-2 | |
| Monoclonal | |
| 0.2mg/mL | |
| GA733-1; TROP2; EGP-1; EGP1; GP50; M1S1; TACD2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 162-46 | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human BeWo (choriocarcinoma) Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.