Learn More
TRIM62 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530999
Description
PA5-30999 targets TRIM62 in IF and WB applications and shows reactivity with Human samples. The PA5-30999 immunogen is recombit fragment corresponding to a region within amino acids 116 and 453 of Human TRIM62. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| TRIM62 | |
| Polyclonal | |
| Unconjugated | |
| Q9BVG3 | |
| TRIM62 | |
| Recombit fragment corresponding to a region within amino acids 116 and 453 of Human TRIM62 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.64 mg/mL | |
| TRIM62 | |
| DEAR1, ductal epithelium-associated RING Chromosome 1, tripartite motif-containing 62, tripartite motif-containing protein 62 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.