Learn More
anti-TRIM24, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00008805-PW1.1Set
Product Code. 16162894
  • £729.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-TRIM24 (300μg), Antibody pair for WB: rabbit polyclonal anti-TRIM24 (50μL)
Specifications
TRIM24
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
PTC6/RNF82/TF1A/TIF1/TIF1A/TIF1ALPHA/hTIF1
Mouse, Rabbit
RUO
8805
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
TRIM24
TRIM24
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-TRIM24 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-TRIM24 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.