missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCHH Monoclonal Antibody (AE15)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA518026

Product Code. 15324301

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This target has a predicted molecular weight of 220 kDa.

This protein is an intermediate filament-associated protein that associates in regular arrays with keratin intermediate filaments (KIF) of the inner root sheath cells of the hair follicle and the granular layer of the epidermis. It later becomes cross-linked to KIF by isodipeptide bonds. It may serve as scaffold protein, together with involucrin, in the organization of the cell envelope or even anchor the cell envelope to the KIF network.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCHH
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
Q07283
TCHH
Urea-mercaptoethanol extract of human hair follicles.
100 μL
Primary
Human, Mouse
Antibody
IgG1 κ
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
AE15
Unconjugated
TCHH
AHF; AI597080; hair follicle protein AHF; Tchh; THH; THL; TRHY; Trichohyalin
Mouse
Protein G
RUO
7062, 99681
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.