missing translation for 'onlineSavingsMsg'
Learn More

anti-TRH, Human, IP-WB Antibody Pair, Abnova™

Artikelnummer. 16192744
Ansicht ändern
Click to view available options
Quantity:
1 Set
Packungsgröße:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16192744 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16192744 Lieferant Abnova Lieferanten-Nr. H00007200PW1.1Set
 Restricted Item
Estimated Shipment: 07-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beinhaltet

Antibody pair for IP: rabbit polyclonal anti-TRH (300μL), Antibody pair for WB: mouse purified polyclonal anti-TRH (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Spezifikation

Antigen TRH
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene TRH
Gene Alias MGC125964/MGC125965
Gene Symbols TRH
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7200
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-TRH (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-TRH (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.