Learn More
TREX1 Mouse anti-Mouse, Unlabeled, Clone: 29, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611987
Description
DNA replication, repair, and recombination requires the excision of nucleotides from the DNA 3′ termini. Many 3′ to 5′ exonucleases have been identified which catalyze the excision of monophosphates from the 3′ termini of DNA. TREX1 and TREX2 are 3′ to 5′ exonucleases that contain three conserved exonuclease active site motifs (EASM) that may produce exonuclease activity. TREX1 and TREX2 are most closely related to the proofreading exonucleases of the bacterial replicative DNA polymerases and the RNase T enzymes. Recombinant expression of TREX1 and TREX2 demonstrates that they have exonuclease activity when oligonucleotide is present. TREX1 shows the greatest exonuclease activity with partial duplex DNA, and no activity with single-stranded RNA or an RNA-DNA partial duplex. In addition, reconstitution of TREX1 with DNA polymerase β and DNA ligase III-XRCC1 facilitates accurate rejoining of a 3′ mismatched base residue at a single-strand break. Thus, TREX1 and TREX2 are 3′ to 5′ exonucleases that may be important for excision of nucleotides during DNA replication, repair, and recombination.
Host Species: Mouse
Clone: 29
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Mouse TRE1 aa. 82-179
Immunofluorescence, Western Blotting
Specifications
| TREX1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse TRE1 aa. 82-179 | |
| 150 μg | |
| Cell Biology | |
| Mouse | |
| IgG1 |
| Western Blot | |
| 29 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.