missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transthyretin Mouse anti-Human, Clone: 10E1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0174
This item is not returnable.
View return policy
Description
A suggested positive control for this product is human plasma.
This gene encodes transthyretin, one of the three prealbumins including alpha-1-antitrypsin, transthyretin and orosomucoid. Transthyretin is a carrier protein; it transports thyroid hormones in the plasma and cerebrospinal fluid, and also transports retinol (vitamin A) in the plasma. The protein consists of a tetramer of identical subunits. More than 80 different mutations in this gene have been reported; most mutations are related to amyloid deposition, affecting predomitly peripheral nerve and/or the heart, and a small portion of the gene mutations is non-amyloidogenic. The diseases caused by mutations include amyloidotic polyneuropathy, euthyroid hyperthyroxinaemia, amyloidotic vitreous opacities, cardiomyopathy, oculoleptomeningeal amyloidosis, meningocerebrovascular amyloidosis, carpal tunnel syndrome, etc.
Specifications
| Transthyretin | |
| Monoclonal | |
| 1 mg/mL | |
| TTR | |
| HsT2651, PALB, TBPA | |
| Mouse | |
| Protein G | |
| RUO | |
| 7276 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 10E1 | |
| Unconjugated | |
| P02766 | |
| TTR | |
| Recombit human protein purified from E.coli | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction