missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM2 Mouse anti-Human, Clone: 2A1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0093
This item is not returnable.
View return policy
Description
A suggested positive control for this product is U87-MG cells.
Transglutaminase (TGase) catalyses the crosslink of proteins by forming e-(g-glutamyl) lysine isopeptide bonds and requires the binding of Ca2+ for its activity. In mammals, eight distinct TGase isoenzymes have been identified. Tissue transglutaminase (tTGase), also known as TGase 2, has four distinct domains: N-terminal b-sandwich, catalytic core and two C-terminal b-barrel domains. tTGase may have a role in cell death, cell proliferation, cell differentiation, and receptor-mediated endocytosis. In the Alzheimer′s disease brain, the elevated tTGase activity is manifested by polymerization of a number of proteins, including Ab peptide, b-amyloid precursor protein and the tau protein, with formation of neurofibrillary tangles.
Specifications
| TGM2 | |
| Monoclonal | |
| 1 mg/mL | |
| TGM2 | |
| C polypeptide, G alpha h, G protein alpha subunit Gh class, | |
| Mouse | |
| Protein G | |
| RUO | |
| 7052 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 2A1 | |
| Unconjugated | |
| P21980 | |
| TGM2 | |
| Purified, recombit, human transglutaminase 2 protein expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction