Learn More
TRA-1-60 Antigen Mouse anti-Human, Unlabeled, Clone: TRA-1-60, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560071
Description
The TRA-1-60 monoclonal antibody reacts with the neuraminidase-resistant form of a pluripotent-stem-cell-specific epitope on a high-molecular-weight transmembrane glycoprotein. The TRA-1-60 antigen is a sialylated epitope on the same keratan sulfate core molecule, podocalyxin, as 4 other distinct antigens on tumor-derived cell lines, TRA-1-81, GCTM2, K4, and K21. The expression of TRA-1-60 antigen is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. The antigen is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (but not morula or trophoblast), and embryonic stem (ES) cells. TRA-1-60 antigen is released into the serum of patients bearing testicular tumors containing EC cells. As human EC and ES cells undergo differentiation, expression of TRA-1-60 antigen is lost. Expression of TRA-1-60 antigen has also been observed on a rhesus monkey ES cell line (Thomson et al, 1995).
Specifications
| TRA-1-60 Antigen | |
| Monoclonal | |
| 0.5mg/mL | |
| TRA-1-60(R) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Rhesus Monkey | |
| IgM κ |
| Western Blot | |
| TRA-1-60 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Embryonal Carcinoma Cell Line | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.