missing translation for 'onlineSavingsMsg'
Learn More
Learn More
torsin family 3, member A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TOR3A protein.
Brand: Abnova H00064222-B01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MLRGPWRQLWLFLLLLLPGAPEPRGASRPWEGTDEPGSAWAWPGFQRLQEQLRAAGALSKRYWTLFSCQVWPDDCDEDEEAATGPLGWRLPLLGQRYLDLLTTWYCSFKDCCPRGDCRISNNFTGLEWDLNVRLHGQHLVQQLVLRTVRGYLETPQPEKALALSFHGWSGTGKNFVARMLVENLYRDGLMSDCVRMFIATFHFPHPKYVDLYKEQLMSQIRETQQLCHQTLFIFDEAEKLHPGLLEVLGPHLERRAPEGHRAESPWTIFLFLSNLRGDIINEVVLKLLKAGWSREEITMEHLEPHLQAEIVETIDNGFGHSRLVKENLIDYFIPFLPLEYRHVRLCARDAFLSQELLYKEETLDEIAQMMVYVPKEEQLFSSQGCKSISQRINYFLSSpecifications
torsin family 3, member A | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TOR3A protein. | |
TOR3A | |
ADIR/ADIR2/FLJ22345/MGC111104 | |
Mouse | |
50 μL | |
Yes | |
64222 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_022371.2 | |
TOR3A | |
TOR3A (AAH11746.1, 1 a.a. ∼ 397 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |