Learn More
CRMP2 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3120
Description
PA3-120 detects TOAD-64 in rat samples. PA3-120 has been successfully used in Western blot, immunoprecipitation and immunohistochemistry (frozen) procedures. By Western blot, this antibody detects an 64 kDa protein representing TOAD-64 in rat samples. The PA3-120 immunogen is a synthetic peptide corresponding to residues Y(499) D G P V F D L T T T P K(511) of TUC-4a (residues 612-624 of TUC-4b).
TOAD-64 is a 64 kDa intracellular neural protein, which belongs to a family of genes that inhibit nerve growth. It also appears to be involved in a role of axon outgrowth. Preliminary results indicate inhibition of TOAD-64 expression poteintiates neurite outgrowth, and may lead to the design of therapies that promote axonal regeneration. Antibodies to TOAD-64 render neurons insensitive to extracellular compounds that inhibit neuron growth. Very few reagents are able to identify newly born neurons as well as antibodies to TOAD-64.
Specifications
| CRMP2 | |
| Polyclonal | |
| Dpysl2 | |
| Turned On After Division 64, DRP2, CRMP-2, DPYL2, Dihydropyrimidinase-related protein 2, Collapsin response mediator protein 2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| O08553, P47942 | |
| DPYSL2 | |
| Synthetic peptide corresponding to residues Y(499) D G P V F D L T T T P K(511) of TUC-4a (residues 612-624 of TUC-4b). | |
| RUO | |
| 12934, 25416 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.