missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNP, PE-CF594, Clone: G155-178, BD
Brand: BD Biosciences 562306
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
| TNP | |
| G155-178 | |
| PE-CF594 | |
| TNP-keyhole limpet hemocyanin | |
| 0.1 mg | |
| Primary |
| Monoclonal | |
| 0.2mg/mL | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction