missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TNFSF9 Matched Antibody Pair, Abnova™
Brand: Abnova H00008744-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-TNFSF9 (100μg); Detection antibody: biotinylated mouse monoclonal anti-TNFSF9, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFSF9.
Host Species: Mouse, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
| TNFSF9 | |
| Unconjugated | |
| 1 Set | |
| Primary |
| Monoclonal | |
| Mouse | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction