missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RII/TNFRSF1B Mouse anti-Human, Clone: 22235, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
Specifications
| Antigen | TNF RII/TNFRSF1B |
|---|---|
| Clone | 22235 |
| Immunogen | E. coli-derived recombinant human TNF RII Extracellular domain |
| Host Species | Mouse |
| Primary or Secondary | Primary |
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| TNF RII/TNFRSF1B | |
| E. coli-derived recombinant human TNF RII Extracellular domain | |
| Primary | |
| Human | |
| Unconjugated | |
| 7133 | |
| Purified | |
| Flow Cytometry, CyTOF | |
| RUO |
| 22235 | |
| Mouse | |
| Monoclonal | |
| IgG2a | |
| CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title