Learn More
TNF-α Mouse anti-Human, FITC, Clone: 6401.1111, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340511
Description
Anti-Hu–TNF-α, clone 6401.1111, is derived from a fusion of Sp2/0 myeloma cells with splenocytes from BALB/c mice immunized with recombinant human TNF-α.
Anti-Human Tumor Necrosis Factor-α (Anti-Hu–TNF-α) recognizes a 26-kilodalton (kd) transmembrane protein.
Specifications
| TNF alpha | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry | |
| 6401.1111 | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Recombinant human TNF-a | |
| 50 Tests | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.