Learn More
TNF Human anti-Mouse/Rat, Biotin, Clone: 516D1A1, BD
Human Monoclonal Antibody
Brand: BD Biosciences 558415
Description
The 516D1A1 Fab antibody reacts with mouse and rat tumor-necrosis factor (TNF, formerly known as TNF-α) proteins. The 516D1A1 Fab was selected by screening a
human Fab phage display library (Hoet et al.) licensed from Dyax Corp (Cambridge, MA) with recombinant mouse TNF protein. The biology and function of the TNF protein has been extensively reviewed (Hehlgans et al.) in the literature.
ELISA
Specifications
| TNF | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse TNF | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| ELISA | |
| 516D1A1 | |
| Biotin | |
| Human | |
| 0.5 mg | |
| Immunology | |
| Mouse, Rat | |
| Ig κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.