missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM59L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542940
This item is not returnable.
View return policy
Description
Peptide sequence: PAASAPSARD PFAPQLGDTQ NCQLRCRDRD LGPQPSQAGL EGASESPYDR Sequence homology: Human: 100%.
This gene encodes a predicted type-I membrane glycoprotein. The encoded protein may play a role in functioning of the central nervous system.
Specifications
| TMEM59L | |
| Polyclonal | |
| Unconjugated | |
| TMEM59L | |
| 5330410G16Rik; brain-specific membrane-anchored protein; BSMAP; C19orf4; RGD1305557; Tmem59l; transmembrane protein 59 like; transmembrane protein 59-like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UK28 | |
| TMEM59L | |
| Synthetic peptide directed towards the N-terminal of human TMEM59L (aa 21-70). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction