Learn More
TMEM126B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543325
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VFRSSLIGIV CGVFYPSSLA FTKNGRLATK YHTVPLPPKG RVLIHWMTLC Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%
TMEM126B Polyclonal antibody specifically detects TMEM126B in Human samples. It is validated for Western Blot.Specifications
TMEM126B | |
Polyclonal | |
Unconjugated | |
TMEM126B | |
1110001A23Rik; complex I assembly factor TMEM126B, mitochondrial; HT007; RGD1308371; TMEM126B; Transmembrane protein 126B | |
Rabbit | |
Protein A | |
RUO | |
55863 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IUX1 | |
TMEM126B | |
Synthetic peptide directed towards the middle region of human TMEM126B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |