missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLL2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534265

Product Code. 15385171

  • £451.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Galago, Marmoset, Rat, Dog, Bat, Pig (100%) Gorilla, Hamster, Panda (92%) Mouse, Horse (85%).

This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. Unlike other family members, a similar protein in mice does not cleave procollagen C-propeptides or chordin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TLL2
Polyclonal
Unconjugated
TLL2
KIAA0932; MTLL-2; TLL2; tolloid like 2; tolloid-like 2; tolloid-like protein 2
Rabbit
Antigen Affinity Chromatography
RUO
100157589, 365460, 486816, 7093
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9Y6L7
TLL2
Synthetic 13 amino acid peptide from CUB 2 domain human TLL2.
50 μg
Primary
Human, Rat, Canine, Monkey, Pig, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.